Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgr5/GPR49 Antibody (803420), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB8240APCCY7
Description
Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Flow CytometrySpecifications
Lgr5/GPR49 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
803420 | |
Flow Cytometry | |
FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
E. coli-derived recombinant mouse Lgr5/GPR49, Lys375-Pro547, Accession # Q9Z1P4 | |
0.1 mL | |
Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway | |
8549 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction