Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lgr5 (GPR49), Mouse, Unlabeled, Clone: 5G10.1,

Catalog No. MABD148MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABD148MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD148MI Supplier MilliporeSigma™ Supplier No. MABD148

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Leucine-rich repeat-containing G-protein coupled receptor 5 (UniProt O75473; also known as G-protein coupled receptor 49, G-protein coupled receptor 67, G-protein coupled receptor HG38, Orphan G protein-coupled receptor HG38) is encoded by the LGR5 (also known as GPR49, GPR67, GRP49, HG38) gene (Gene ID 8549) in human. Lgr5 is a 7-transmembrane protein related to the receptors for the follicle stimulating hormone (FSH), the luteinizing hormone (LH), and the thyroid-stimulating hormone (TSH). Lgr5 is a Wnt signaling pathway targeted gene expressed in proliferative stem cells in various tissues, including small intestine, colon, stomach, and the hair follicle. The R-spondin (RSPO) family of secreted Wnt signaling agonists are known to bind Lgr5 and the related Lgr4 and Lgr6, and RSPO1 depends on its association wth Lgr4, Lgr5, or Lgr6 for its Wnt signaling enhancing effect. Human Lgr5 is synthesized with a propeptide sequence (a.a. 1-21), the removal of which yields the mature 7-transmembrane (a.a. 562-582 , 594-614, 639-659, 683-703, 723-743, 768-788, 803-823) G-protein-coupled receptor (GPCR), having its N-terminal end (a.a. 22-561) at the extracellular side and its C-terminal end (a.a. 824-907) at the cytoplasmic side. The N-terminal extracellular region contains a leucine-rich repeat (LRR) N-terminal (LRRNT) domain (a.a. 25-66), followed by 16 LRRs (LRR 1 through LLR 16; a.a. 67-446).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgr5 (GPR49)
Applications Western Blot
Classification Monoclonal
Clone 5G10.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene LGR5, GPR49, GPR67, GRP49, HG38
Gene Accession No. O75473
Host Species Mouse
Immunogen GST-tagged recombinant human Lgr5/GPR49 fragment corresponding to leucine-rich repeats (LRRs) 7-11 within the N-terminal extracellular domain.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Research
Primary or Secondary Primary
Gene ID (Entrez) NP_003658
Test Specificity Clone 5G10.1 targets an epitope within human Lgr5 (GPR49) N-terminal extracellular region comprised of leucine-rich repeats (LRRs) 7-11. Reactivity toward spliced isoform 2 that lacks part of the LLRs 9&10 sequence is possible, but has not been confirmed.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.