Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGR5 Monoclonal Antibody (GT11312)
GREENER_CHOICE

Catalog No. PIMA531523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531523 Supplier Invitrogen™ Supplier No. MA531523
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (84%), Rat (83%), Pig (89%), Bovine (86%). Positive Control: unboiled Myc-tagged hLGR5-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

G protein-coupled receptor GPR49 has been reported to be expressed in brain, skeletal muscle, placenta, and spinal cord. ESTs have been isolated from normal human brain (amygdala), embryo, and placenta libraries and from human germ cell, uterus, and brain libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This 'turn-off' activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGR5
Applications Western Blot
Classification Monoclonal
Clone GT11312
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative
Gene Lgr5
Gene Accession No. O75473, Q9Z1P4
Gene Alias Fex; follicle expressed hormone; G protein-coupled receptor 49; GPR49; GPR67; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; GRP49; HG38; leucine rich repeat containing G protein coupled receptor 5; leucine rich repeat containing G protein-coupled receptor 5; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; leucine-rich repeat-containing G-protein coupled receptor 5; Lgr5; orphan G protein-coupled receptor HG38; orphan G-protein coupled receptor FEX
Gene Symbols Lgr5
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human LGR5. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14160, 8549
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.