Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LGR5 Mouse anti-Human, Mouse, DyLight™ 550, Clone: OTI2A2, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA400003

Description
G protein-coupled receptor GPR49 has been reported to be expressed in brain, skeletal muscle, placenta, and spinal cord. ESTs have been isolated from normal human brain (amygdala), embryo, and placenta libraries and from human germ cell, uterus, and brain libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This ""turn-off"" activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.Specifications
LGR5 | |
Monoclonal | |
0.73 mg/mL | |
PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide | |
O75473, Q9Z1P4 | |
LGR5 | |
Human recombit protein fragment corresponding to amino acids 250-550 of human LGR5 produced in HEK293T Cell. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence | |
OTI2A2 | |
DyLight 550 | |
LGR5 | |
FEX, GPR49, GPR67, GRP49, HG38 | |
Mouse | |
Affinity Chromatography | |
RUO | |
14160, 8549 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction