Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr5 Rat anti-Human, Brilliant Violet 605, Clone: 8F2, BD Optibuild™

Catalog No. BDB742887 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742887 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742887 Supplier BD Biosciences Supplier No. 742887
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Lgr5 (leucine-rich-repeat-containing G-protein-coupled receptor 5) is a seven transmembrane-domain receptor that is a target gene for Wnt and marks stem cells in the small intestine, colon, stomach, and hair follicle. Lgr5 was initially identified as a potential stem cell marker due to restricted expression of Lgr5 in the intestinal crypt and labeling of rapidly cycling cells of the colon and intestine. Using both lineage tracing and organoid culture experiments, Lgr5 positive cells are capable of generating all types of the small intestine epithelium hence indicating that Lgr5 marks stem cells of the small intestine and colon. R-spondin growth factors, which are secreted agonists of the Wnt pathway, bind Lgr5. The binding of R-spondins to Lgr5 leads to recruitment of the Frizzled/LRP Wnt receptor complex, which binds to Wnt ligands and leads to downstream Wnt signaling. Lgr5 is up-regulated in colon and ovarian cancers and has been implicated in promotion of tumor growth and metastasis.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgr5
Classification Monoclonal
Clone 8F2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias GPR49, GPR67, HG38
Host Species Rat
Immunogen Human LGR5
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.