Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgr5 Rat anti-Human, BUV496, Clone: 8F2, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 750279
Description
Lgr5 Monoclonal antibody specifically detects Lgr5 in Human samples. It is validated for Flow Cytometry
Specifications
Lgr5 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
GPR49, GPR67, HG38 | |
Human LGR5 | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
8F2 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction