Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgr6 Antibody (OTI2A3), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272119AF350
Description
Lgr6 Monoclonal specifically detects Lgr6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Lgr6 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
LGR6 | |
Recombinant protein fragment corresponding to amino acids 25-250 of human Lgr6 (NP_001017403) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2A3 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
Mouse | |
Affinity Purified | |
GPCR | |
59352.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction