Learn More
Invitrogen™ LGR6 Recombinant Rabbit Monoclonal Antibody (8J2J10)

Description
Immunogen sequence: FPVTPEAVKS VLLVVLPLPA CLNPLLYLLF NPHFRDDLRR LRPRAGDSGP LAYAAAGELE KSSCDSTQAL VAFSDVDLIL EASEAGRPPG LETYGFPSVT.
Specifications
Specifications
| Antigen | LGR6 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 8J2J10 |
| Concentration | 0.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Lgr6 |
| Gene Accession No. | Q9HBX8 |
| Gene Alias | A530037C04Rik; D830026M09; FLJ14471; gonadotropin receptor; GPCR; leucine rich repeat containing G protein-coupled receptor 6; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; leucine-rich repeat-containing G-protein coupled receptor 6; Lgr6; RGD1560339; RP11-572A16.3; UNQ6427/PRO21331; VTS20631 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.