Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LGR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102099
Description
Antibody detects endogenous levels of total LGR6.
LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.
Specifications
| LGR6 | |
| Polyclonal | |
| Unconjugated | |
| Lgr6 | |
| A530037C04Rik; D830026M09; FLJ14471; gonadotropin receptor; GPCR; leucine rich repeat containing G protein-coupled receptor 6; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; leucine-rich repeat-containing G-protein coupled receptor 6; Lgr6; RGD1560339; RP11-572A16.3; UNQ6427/PRO21331; VTS20631 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 329252, 59352 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UVD5, Q9HBX8 | |
| Lgr6 | |
| A synthesized peptide derived from human LGR6(Accession Q9HBX8), corresponding to amino acid residues Q478-D528. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction