Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598995
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598995 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598995 Supplier Invitrogen™ Supplier No. PA598995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LGR6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGR6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Lgr6
Gene Accession No. Q9HBX8
Gene Alias A530037C04Rik; D830026M09; FLJ14471; gonadotropin receptor; GPCR; leucine rich repeat containing G protein-coupled receptor 6; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; leucine-rich repeat-containing G-protein coupled receptor 6; Lgr6; RGD1560339; RP11-572A16.3; UNQ6427/PRO21331; VTS20631
Gene Symbols Lgr6
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich repeat-containing G-protein coupled receptor 6 protein (839-962AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59352
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.