Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599926 Supplier Invitrogen™ Supplier No. PA599926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RXFP1.

LGR7 is a low density lipoprotein receptor of 757amino acid belonging to the G-protein coupled receptor 1 family with rhodopsin-like receptor activity. It has seven transmembrane region, cysteine-rich motif, 10 LRR domains and 5 N-glycosylation sites in the N terminus. Research reports show that it has receptor for relaxins (RLN1 and RLN2) which stimulates adenylate cyclase activation, substrate adhesion, and chemo migratory capacity of mononuclear leukocytes. LGR7 is critical for mediating these biological responses by use of RNA interference lentiviral short hairpin constructs. It is widely expressed in 86% of 15 primary EC tissue samples including brain, kidney, testis and endometrium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGR7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RXFP1
Gene Accession No. Q6R6I6, Q6R6I7, Q9HBX9
Gene Alias Gm1018; leucine-rich repeat-containing 7; leucine-rich repeat-containing G protein-coupled receptor 7; leucine-rich repeat-containing G-protein coupled receptor 7; LGR7; LGR7.1; LGR7.10; LGR7.2; MGC138347; MGC142177; Relaxin family peptide receptor 1; relaxin receptor 1; relaxin/insulin like family peptide receptor 1; relaxin/insulin-like family peptide receptor 1; RXFP1; RXFPR1
Gene Symbols RXFP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RXFP1(Accession Q9HBX9), corresponding to amino acid residues R527-Q577.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295144, 381489, 59350
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.