Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGSN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565252 Supplier Invitrogen™ Supplier No. PA565252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSRSKTIPAHF FQEKVSHGVC MPRGYLEVIP NPKDNEMNNI RATCFNSDIV LMPELSTFRV LPWADRTARV ICDTFTVTGE PLLTSPRY Highest antigen sequence identity to the following orthologs - mouse 90%, rat 90%.

May act as a component of the cytoskeleton or as a chaperone for the reorganization of intermediate filament proteins during terminal differentiation in the lens. Does not seem to have enzymatic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGSN
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LGSN
Gene Accession No. Q5TDP6
Gene Alias GLULD1; glutamate-ammonia ligase (glutamine synthase) domain containing 1; glutamate-ammonia ligase domain-containing protein 1; Lengsin; lengsin, lens protein with glutamine synthetase domain; Lens glutamine synthase-like; LGS; LGSN
Gene Symbols LGSN
Host Species Rabbit
Immunogen Recombinant Human LGSN. Recombinant protein control fragment (Product #RP-104711).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51557
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.