Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LGTN Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150038UV
Description
LGTN Polyclonal antibody specifically detects LGTN in Human samples. It is validated for Western BlotSpecifications
LGTN | |
Polyclonal | |
Western Blot | |
eukaryotic translation initiation factor 2D, HCA56eIF2d, Hepatocellular carcinoma-associated antigen 56, LGTNLigatin, ligatin | |
The immunogen for this product maps to a region between residue 350 and 400 of human eIF2D using the numbering given in entry NP_008824.2 (GeneID 1939). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
1939 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction