Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGTN Antibody, Novus Biologicals™
SDP

Catalog No. p-7107448 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP157395 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP157395 Supplier Novus Biologicals Supplier No. NBP157395
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LGTN Polyclonal specifically detects LGTN in Human samples. It is validated for Western Blot.

Specifications

Antigen LGTN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P41214
Gene Alias eukaryotic translation initiation factor 2D, HCA56eIF2d, Hepatocellular carcinoma-associated antigen 56, LGTNLigatin, ligatin
Gene Symbols EIF2D
Host Species Rabbit
Immunogen Synthetic peptides corresponding to eIF2D. The peptide sequence was selected from the middle region of eIF2D. Peptide sequence KVTVVRNLEAYGLDPYSVAAILQQRCQASTTVNPAPGAKDSLQVQIQGNQ.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1939
Test Specificity Expected identity based on immunogen sequence: Sumatran orangutan: 100%; Human: 100%; Canine: 92%; Rat: 85%; Rhesus macaque: 85%; Mouse: 85%;.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.