Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LH beta Antibody (L1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073010IR
Description
LH beta Monoclonal specifically detects LH beta in Human samples. It is validated for Western Blot, Sandwich ELISA.Specifications
LH beta | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LHB | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified natural LH beta. | |
0.2 mL | |
Primary | |
High affinity for the beta chain of LH. No cross-reaction with FSH, TSH or HCG. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
L1 | |
Western Blot, Sandwich ELISA | |
CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta | |
Mouse | |
Protein A or G purified | |
RUO | |
3972 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction