Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LH beta Antibody (SPM103), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311530UV
Description
LH beta Monoclonal antibody specifically detects LH beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
LH beta | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
3972 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SPM103 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta | |
Recombinant beta sub-unit of human LH beta (Uniprot: P01229) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction