Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LH beta Mouse anti-Human, Clone: rLHb/1613, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP31395920UG
This item is not returnable.
View return policy
Description
LH beta Monoclonal antibody specifically detects LH beta in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
LH beta | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
P01229 | |
Mouse | |
Protein A or G purified | |
RUO | |
3972 | |
Store at 4°C. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rLHb/1613 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta | |
Recombinant full-length human LH beta protein (Uniprot: P01229) | |
20 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction