Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LHFPL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551800
Description
Immunogen sequence: GKPVSFSTFR RCNYPVRGEG HSLIMVEECG RYASFNAIPS LAWQMCT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in one LHFP-like gene result in deafness in humans and mice, and a second LHFP-like gene is fused to a high-mobility group gene in a translocation-associated lipoma. Alternatively spliced transcript variants have been found, but their biological validity has not been determined.
Specifications
| LHFPL1 | |
| Polyclonal | |
| Unconjugated | |
| LHFPL1 | |
| LHFPL tetraspan subfamily member 1 protein; LHFPL1; LHFP-like protein 1; lipoma HMGIC fusion partner-like 1; lipoma HMGIC fusion partner-like 1 protein; UNQ5824/PRO19643 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 340596 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86WI0 | |
| LHFPL1 | |
| Recombinant protein corresponding to Human LHFPL1. Recombinant protein control fragment (Product #RP-92254). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction