Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHFPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559967 Supplier Invitrogen™ Supplier No. PA559967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAGWGCQKAI DYCGHYASAY KPGDC Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

LHPL2 is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHFPL2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LHFPL2
Gene Accession No. Q6ZUX7
Gene Alias 6030465B15; AI447312; AW050335; I79_004617; KIAA0206; LHFPL tetraspan subfamily member 2 protein; Lhfpl2; LHFP-like protein 2; lipoma HMGIC fusion partner-like 2; lipoma HMGIC fusion partner-like 2 protein; mKIAA0206; vgim
Gene Symbols LHFPL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LHFPL2. Recombinant protein control fragment (Product #RP-91158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.