Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LHFPL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567063
Description
Immunogen sequence: VLGNRQDSLMA EELKAENKVL LSQYSLE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 50%.
LHFPL3 is a member of the lipoma HMGIC fusion partner(LHFP) gene family, which is a subset of the superfamily oftetraspan transmembrane protein encoding genes. Mutations in oneLHFP-like gene result in deafness in humans and mice, and a secondLHFP-like gene is fused to a high-mobility group gene in atranslocation-associated lipoma. A partial gene fragment namedLHFPL4 corresponds to a portion of the first exon of this gene.
Specifications
LHFPL3 | |
Polyclonal | |
Unconjugated | |
LHFPL3 | |
A930031L14Rik; GABAA receptor regulatory Lhfpl4; GARLH4; LHFPL tetraspan subfamily member 3 protein; LHFPL tetraspan subfamily member 4 protein; LHFPL3; LHFPL4; LHFP-like protein 3; LHFP-like protein 4; lipoma HMGIC fusion partner-like 3; lipoma HMGIC fusion partner-like 3 protein; lipoma HMGIC fusion partner-like 4; lipoma HMGIC fusion partner-like 4 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375612 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UP9 | |
LHFPL3 | |
Recombinant Human LHFPL3. Recombinant protein control fragment (Product #RP-107703). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction