Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHFPL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570831 Supplier Invitrogen™ Supplier No. PA570831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

LHFPL4 plays a role in the regulation of inhibitory synapse formation and function by being involved in maintaining gamma-aminobutyric acid receptors (GABAARs) clustering and their associated scaffold proteins at inhibitory synaptic sites. Acts in concert with NLGN2 to recruit or stabilize GABAARs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHFPL4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LHFPL4
Gene Accession No. Q7Z7J7
Gene Alias 1190004M23Rik; AI604880; B230384L07; GABAA receptor regulatory Lhfpl4; Garlh4; HMGIC fusion partner-like protein 4; Kiaa4027; Lh4; LHFPL tetraspan subfamily member 4; LHFPL tetraspan subfamily member 4 protein; LHFPL3; Lhfpl4; LHFP-like protein 4; lipoma HMGIC fusion partner-like 3; lipoma HMGIC fusion partner-like 4; lipoma HMGIC fusion partner-like 4 protein; lipoma HMGIC fusion partner-like protein 4; mKIAA4027
Gene Symbols LHFPL4
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human LHFPL4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375323
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.