Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHFPL5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP324750
Description
LHFPL5 Polyclonal antibody specifically detects LHFPL5 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
LHFPL5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
DFNB67, dJ510O8.8, LHFP-like protein 5, lipoma HMGIC fusion partner-like 5, Lipoma HMGIC fusion partner-like 5 protein, MGC33835, tetraspan membrane protein of hair cell stereocilia, Tmhs, TMHSdeafness, autosomal recessive 67 | |
This antibody has been engineered to specifically recognize the recombinant protein LHFPL5 using the following amino acid sequence: GDSVNTPQAGYFGLFSYCVGNVLSSELICKGGPLDFSSIPS | |
100 μL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
222662 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction