Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHFPL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144747 Supplier Invitrogen™ Supplier No. PA5144747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDSVNTPQAG YFGLFSYCVG NVLSSELICK GGPLDFSSIP S Antigen sequence identity: Mouse 98%; Rat 98%.

This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in this gene result in deafness in humans, and a mutation in a similar gene in mice results in deafness and vestibular dysfunction with severe degeneration of the organ of Corti. It is proposed to function in hair bundle morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHFPL5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LHFPL5
Gene Accession No. Q8TAF8
Gene Alias 9330179O15Rik; DFNB67; dJ510O8.8; hscy; LHFPL tetraspan subfamily member 5 protein; Lhfpl5; LHFP-like protein 5; lipoma HMGIC fusion partner-like 5; lipoma HMGIC fusion partner-like 5 protein; Tetraspan membrane protein of hair cell stereocilia; tetraspan transmembrane protein, hair cell stereocilia; TMHS
Gene Symbols LHFPL5
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 51-91. Recombinant protein control fragment (Product #RP-109978).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.