Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHR Antibody (LHCGR/1415), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254494PEATT594
Description
LHR Monoclonal specifically detects LHR in Human samples. It is validated for Flow Cytometry.Specifications
LHR | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
LHCGR | |
Recombinant fragment (around aa 70-410) of human LHR protein (exact sequence is proprietary) (Uniprot: P22888) | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
LHCGR/1415 | |
Flow Cytometry | |
FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor | |
Mouse | |
Protein A or G purified | |
GPCR | |
3973.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction