Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHR Antibody (LHCGR/1417) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | LHR |
---|---|
Clone | LHCGR/1417 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254479100
![]() |
Novus Biologicals
NBP254479100UG |
100 μg |
Each for $646.00
|
|
|||||
NB254479200
![]() |
Novus Biologicals
NBP254479200UG |
200 μg | N/A | N/A | N/A | ||||
Description
LHR Monoclonal specifically detects LHR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
LHR | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
GPCR | |
PBS with No Preservative | |
3973 | |
Recombinant N-terminal fragment of human LHCGR (exact sequence is proprietary) | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
LHCGR/1417 | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor | |
LHCGR | |
IgG1 κ | |
Protein A or G purified | |
82 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title