Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHR Antibody (LHCGR/1417), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB281084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB281084 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB281084 Supplier Novus Biologicals Supplier No. NBP254479MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LHR Monoclonal antibody specifically detects LHR in Human samples. It is validated for Flow Cytometry

Specifications

Antigen LHR
Applications Flow Cytometry
Classification Monoclonal
Clone LHCGR/1417
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor
Host Species Mouse
Immunogen Recombinant fragment (around aa 70-410) of human LHR protein (exact sequence is proprietary) (Uniprot: P22888)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Caveolae Markers, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3973
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.