Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHR Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP276804
Description
LHR Polyclonal specifically detects LHR in Human samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA, Immunofluorescence.Specifications
LHR | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor | |
Goat | |
Affinity Purified | |
Caveolae Markers, GPCR | |
3973.0 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
0.50 mg/ml | |
Western Blot 0.3 - 0.1 ug/ml, Flow Cytometry 10 ug/ml, Peptide ELISA Detection Limit 1:32000, Immunofluorescence 10 ug/ml | |
P22888, NP_000224.2 | |
LHCGR | |
Peptide with sequence CQGTALLDKTRYTE corresponding to C-Terminus according to NP_000224.2. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction