Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533836
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533836 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533836 Supplier Invitrogen™ Supplier No. PA533836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Marmoset, Rat, Dog, Panda (95%) Bovine, Bat, Rabbit, Horse (89%) Ferret (84%).

LHCGR encodes the receptor for both luteinizing hormone and choriogonadotropin. It is family 1 of G-protein coupled receptors, FSH/LSH/TSH superfamily, and its basic function is acting as luteinizing hormone/choriogonadotropin receptor (gonadotropin receptor). Regulating the development and function of Leyding cells in males. It also plays a role in male secondary sexual characterdevelopment. Mutations in this gonadotropinreceptor results in disorders of male secondary sexual characteristic development, such as familial male precocious puperty and male pseudohermaphtoditism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LHCGR
Gene Accession No. P22888
Gene Alias Gpcr19-rs1; HCG receptor; HHG; hypergonadotropic hypogonadism; LCGR; LGR2; LH receptor; LH/CGR; LH/CG-R; LHCGR; LHR; LH-R; LHRHR; LSHR; LSH-R; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; Lutropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; ULG5
Gene Symbols LHCGR
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human hCG receptor.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3973
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.