Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597923
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597923 Supplier Invitrogen™ Supplier No. PA597923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LHR Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

LHCGR encodes the receptor for both luteinizing hormone and choriogonadotropin. It is family 1 of G-protein coupled receptors, FSH/LSH/TSH superfamily, and its basic function is acting as luteinizing hormone/choriogonadotropin receptor (gonadotropin receptor). Regulating the development and function of Leyding cells in males. It also plays a role in male secondary sexual characterdevelopment. Mutations in this gonadotropinreceptor results in disorders of male secondary sexual characteristic development, such as familial male precocious puperty and male pseudohermaphtoditism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LHCGR
Gene Accession No. P22888, P30730
Gene Alias Gpcr19-rs1; HCG receptor; HHG; hypergonadotropic hypogonadism; LCGR; LGR2; LH receptor; LH/CGR; LH/CG-R; LHCGR; LHR; LH-R; LHRHR; LSHR; LSH-R; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; Lutropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; ULG5
Gene Symbols LHCGR
Host Species Rabbit
Immunogen Recombinant Human Lutropin-choriogonadotropic hormone receptor protein (27-363AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16867, 3973
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.