Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHX1 Monoclonal Antibody (OTI1D5), TrueMAB™, OriGene
SDP

Catalog No. 502540269
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0269 30 μL
50-167-5197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0269 Supplier OriGene Supplier No. TA504533S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LHX1 Monoclonal antibody specifically detects LHX1 in Human samples. It is validated for Western Blot

Specifications

Antigen LHX1
Applications Western Blot
Classification Monoclonal
Clone OTI1D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Lhx1
Gene Accession No. P48742
Gene Alias hLim-1; Homeobox protein Lim-1; LHX1; LHX-1; LIM homeo box protein 1; LIM homeobox 1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Lim1; Lim-1; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx1; MGC126723; MGC138141; rlim; transcription factor; homeobox protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 100-362 of human LHX1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3975
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.