Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHX1 Monoclonal Antibody (OTI2A4), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA504528

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-5192


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of neural and lymphoid cells. A similar protein in mice is an essential regulator of the vertebrate head organizer.
Specifications

Specifications

LHX1
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P48742
LHX1
Human recombit protein fragment corresponding to amino acids 100-362 of human LHX1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2A4
Unconjugated
LHX1
LIM-1, LIM1
Mouse
Affinity Chromatography
RUO
3975
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.