Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHX1 Monoclonal Antibody (OTI2A4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504528S


Description
LHX1 Monoclonal antibody specifically detects LHX1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
LHX1 | |
Monoclonal | |
0.69 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P48742 | |
Mouse | |
Affinity chromatography | |
RUO | |
3975 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2A4 | |
Unconjugated | |
Lhx1 | |
hLim-1; Homeobox protein Lim-1; LHX1; LHX-1; LIM homeo box protein 1; LIM homeobox 1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Lim1; Lim-1; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx1; MGC126723; MGC138141; rlim; transcription factor; homeobox protein | |
Human recombinant protein fragment corresponding to amino acids 100-362 of human LHX1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction