Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHX2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$336.16 - $678.13
Specifications
| Antigen | LHX2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
LHX2 Polyclonal specifically detects LHX2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| LHX2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| hLhx2, Homeobox protein LH-2, LH-2, LH2MGC138390, LIM homeobox 2, LIM homeobox protein 2, LIM HOX gene 2, LIM/homeobox protein Lhx2 | |
| LHX2 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 9355 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGSAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYN | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title