Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHX2 Antibody (SR1750), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32237150UL
Description
LHX2 Monoclonal antibody specifically detects LHX2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
LHX2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1750 | |
Western Blot 1:500-1:2000 | |
hLhx2, Homeobox protein LH-2, LH-2, LH2MGC138390, LIM homeobox 2, LIM homeobox protein 2, LIM HOX gene 2, LIM/homeobox protein Lhx2 | |
A synthesized peptide derived from human LHX2 (Uniprot #: P50458) | |
50 μg | |
Neuroscience, Stem Cells | |
9355 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction