Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX2 Monoclonal Antibody (6G2)
GREENER_CHOICE

Catalog No. PIMA515834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515834 Supplier Invitrogen™ Supplier No. MA515834
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15834 targets LHX2 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15834 immunogen is purified recombinant fragment of human LHX2 expressed in E. Coli. . MA5-15834 detects LHX2 which has a predicted molecular weight of approximately 44kDa.

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6G2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Lhx2
Gene Accession No. P50458
Gene Alias ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390
Gene Symbols Lhx2
Host Species Mouse
Immunogen Purified recombinant fragment of human LHX2 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9355
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.