Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LHX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568971
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications
| LHX2 | |
| Polyclonal | |
| Unconjugated | |
| Lhx2 | |
| ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16870 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Z0S2 | |
| Lhx2 | |
| synthetic peptide directed towards the C-terminal region of mouse LHX2. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction