Learn More
Invitrogen™ LHX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578287
Description
Positive Control: 293T, A431, HeLa, HepG2, Neuro 2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, mouse brain, mouse eye Predicted Reactivity: Pig (98%), Chicken (81%), Rhesus Monkey (98%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications
LHX2 | |
Polyclonal | |
Unconjugated | |
Lhx2 | |
ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16870, 296706, 9355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
0.97 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P36198, P50458, Q9Z0S2 | |
Lhx2 | |
Recombinant protein encompassing a sequence within the C-terminus region of human LHX2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Fish | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.