Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540754 Supplier Invitrogen™ Supplier No. PA540754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QNRRAKEKRL KKDAGRQRWG QYFRNMKRSR GGSKSDKDSV QEGQDSDAEV Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 87%; Human: 100%; Mouse: 93%; Rat: 100%; Zebrafish: 80%.

This gene encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine. Two transcripts variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LHX3
Gene Accession No. Q9UBR4
Gene Alias bLhx3b; CPHD3; DKFZp762A2013; Homeobox protein LIM-3; homeobox protein P-LIM; lhx3; Lhx3b; LIM homeo box protein 3; LIM homeobox 3; LIM homeobox protein; LIM homeobox protein 3; LIM homeodomain 3; LIM homeodomain 3 protein b isoform; LIM homeodomain box protein 3; LIM homeodomain protein 3b; LIM/homeobox protein Lhx3; LIM/homeodomain protein LHX3; lim3; Lim-3; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx3; M2-LHX3; mLIM3; mLim-3; Plim; P-LIM
Gene Symbols LHX3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LHX3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8022
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.