Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598891
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598891 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598891 Supplier Invitrogen™ Supplier No. PA598891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LHX3 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine. Two transcripts variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LHX3
Gene Accession No. Q9UBR4
Gene Alias bLhx3b; CPHD3; DKFZp762A2013; Homeobox protein LIM-3; homeobox protein P-LIM; lhx3; Lhx3b; LIM homeo box protein 3; LIM homeobox 3; LIM homeobox protein; LIM homeobox protein 3; LIM homeodomain 3; LIM homeodomain 3 protein b isoform; LIM homeodomain box protein 3; LIM homeodomain protein 3b; LIM/homeobox protein Lhx3; LIM/homeodomain protein LHX3; lim3; Lim-3; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx3; M2-LHX3; mLIM3; mLim-3; Plim; P-LIM
Gene Symbols LHX3
Host Species Rabbit
Immunogen Recombinant Human LIM/homeobox protein Lhx3 protein (233-389AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8022
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.