Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117116 Supplier Invitrogen™ Supplier No. PA5117116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Rat brain Immunogen sequence: NRQLATGDEF YLMEDGRLVC KEDYETAKQN DDSEAGAKRP RTTITAKQLE TLKNAYKNSP KPARHVREQL SSETGLDMRV VQVWFQNRRA KEKRLKKDAG RHRWGQFYKS VKRSRGSSKQ EKESSAEDCG VSDSELSFRE DQILSELGHT NRIYGNVGDV TGGQLMNGSF SMDGTGQSYQ DLRDGSPYGI PQSPSSISSL PSHAPLLNGL DYTVDSNLGI IAHAGQGVSQ TLRAMAGGPT SDISTGSSVG YPDFPTSPGS WLDEMDHPPF.

This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of the pituitary gland. Mutations in this gene are associated with syndromic short stature and pituitary and hindbrain defects. An alternative splice variant has been described but its biological nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LHX4
Gene Accession No. P53776, Q969G2
Gene Alias A330062J17Rik; CPHD4; Gsh4; Gsh-4; Lhx4; LIM homeo box protein 4; LIM homeobox 4; LIM homeobox protein 4; LIM/homeobox protein Lhx4; LXH4
Gene Symbols LHX4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 121-390 of human LHX4 (NP_2031291).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16872, 360858, 89884
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.