Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHX5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24910325UL
Description
LHX5 Polyclonal antibody specifically detects LHX5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
LHX5 | |
Polyclonal | |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
LIM homeobox 5, LIM homeobox protein 5, LIM/homeobox protein Lhx5, MGC129689 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LQDALQDDPKETDNSTSSDKETANNENEEQNSGTK | |
25 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
64211 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction