Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX5 Monoclonal Antibody (2A8)
GREENER_CHOICE

Catalog No. PIMA522476
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522476 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522476 Supplier Invitrogen™ Supplier No. MA522476
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: CTDRSLSPDL QDALQDDPKE TDNSTSSDKE TANNENEEQN SGTKRRGPRT TIKAKQLETL KAAFAATPKP TRHIREQLAQ ETGLNMRVIQ VWFQNRRSKE.

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in the control of differentiation and development of the forebrain. In mice, this protein is essential for the regulation of precursor cell proliferation and the control of neuronal differentiation and migration during hippocampal development. This protein is involved in learning and motor functions in adult mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX5
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2A8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene LHX5
Gene Accession No. Q9H2C1
Gene Alias homeobox protein LIM-2; Homeobox protein LIM-5; Lhx5; LIM class homeodomain protein; LIM homeo box protein 5; LIM homeobox 5; LIM homeobox protein 5; LIM/homeobox protein Lhx5; Lim2; Lim-2; lim5; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx5; MGC129689
Gene Symbols LHX5
Host Species Mouse
Immunogen LHX5 (NP_071758, 136 a.a. ~ 235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64211
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.