Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547828
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547828 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547828 Supplier Invitrogen™ Supplier No. PA547828
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody detects recombinant human LHX5 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) LIM1 is observed, and less than 5% cross-reactivity with rhLHX2, and rhLHX9 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in the control of differentiation and development of the forebrain. In mice, this protein is essential for the regulation of precursor cell proliferation and the control of neuronal differentiation and migration during hippocampal development. This protein is involved in learning and motor functions in adult mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX5
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LHX5
Gene Accession No. P61375, P61376, Q9H2C1
Gene Alias homeobox protein LIM-2; Homeobox protein LIM-5; Lhx5; LIM class homeodomain protein; LIM homeo box protein 5; LIM homeobox 5; LIM homeobox protein 5; LIM/homeobox protein Lhx5; Lim2; Lim-2; lim5; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx5; MGC129689
Gene Symbols LHX5
Host Species Goat
Immunogen E. coli-derived recombinant human LHX5 Leu265-Trp402.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124451, 16873, 64211
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.