Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540560 Supplier Invitrogen™ Supplier No. PA540560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LSRRVIQVWF QNCRARHKKH TPQHPVPPSG APPSRLPSAL SDDIHYTPFS Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Rat: 100%.

LHX6 (LIM/homeobox protein Lhx6) is a probably transcription factor required for the expression of a subset of genes involved in interneurons migration and development. It functions in the specification of cortical interneuron subtypes and in the migration of GABAergic interneuron prescursors from the subpallium to the cerebral cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LHX6
Gene Accession No. Q9UPM6
Gene Alias LHX6; LHX6.1; LIM homeobox 6; LIM homeobox protein 6; LIM homeodomain protein 6.1; LIM/homeobox protein Lhx6; LIM/homeobox protein Lhx6.1; LIM-homeodomain protein 6.1; MGC119542; MGC119544; MGC119545; RP11-498E2.6
Gene Symbols LHX6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LHX6 (aa 258-307).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26468
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.