Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LHX6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115509
Description
Antibody detects endogenous levels of total LHX6.
LHX6 (LIM/homeobox protein Lhx6) is a probably transcription factor required for the expression of a subset of genes involved in interneurons migration and development. It functions in the specification of cortical interneuron subtypes and in the migration of GABAergic interneuron prescursors from the subpallium to the cerebral cortex.
Specifications
LHX6 | |
Polyclonal | |
Unconjugated | |
LHX6 | |
LHX6; LHX6.1; LIM homeobox 6; LIM homeobox protein 6; LIM homeodomain protein 6.1; LIM/homeobox protein Lhx6; LIM/homeobox protein Lhx6.1; LIM-homeodomain protein 6.1; MGC119542; MGC119544; MGC119545; RP11-498E2.6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16874, 26468, 311901 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q9R1R0, Q9UPM6 | |
LHX6 | |
A synthesized peptide derived from human LHX6(Accession Q9UPM6), corresponding to amino acid residues T256-F306. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction