Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LHX8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531927 Supplier Invitrogen™ Supplier No. PA531927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (94%), Pig (100%), Chicken (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the LIM homeobox gene family, such as LHX8, encode transcription regulators that share common structural features. They all contain 2 tandemly repeated cysteine-rich double-zinc finger motifs, called LIM domains, in addition to a homeodomain. The homeodomain is a DNA-binding domain, and the LIM domains are essential for regulating the activity of these molecules by interacting with other proteins. Members of the LIM homeobox gene family are required for the patterning or the specification and differentiation of different cell types during embryonic development (Zhao et al., 1999 [PubMed 10611327]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX8
Applications Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene LHX8
Gene Accession No. Q68G74
Gene Alias L3; Lhx7; LHX8; LIM homeo box protein 8; LIM homeobox 8; LIM homeobox protein 7; LIM homeobox protein 8; LIM Homeodomain 8; LIM/homeobox protein Lhx7; LIM/homeobox protein Lhx8; LIM-homeodomain protein Lhx8
Gene Symbols LHX8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LHX8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 431707
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.