Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LHX9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541160
Description
Peptide sequence: RNLLRQENGG VDKADGTSLP APPSADSGAL TPPGTATTLT DLTNPTITVV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 100%.
LHX9 encodes a member of the LIM homeobox gene family of developmentally expressed transcription factors. The encoded protein contains a homeodomain and two cysteine-rich zinc-binding LIM domains involved in protein-protein interactions. The protein is highly similar to a mouse protein that causes gonadal agenesis when inactivated, suggesting a role in gonadal development.
Specifications
LHX9 | |
Polyclonal | |
Unconjugated | |
LHX9 | |
3110009O07Rik; BB104635; LH2B; Lhx9; LIM homeobox 9; LIM homeobox 9-like; LIM homeobox protein 9; LIM homeodomain type transcription factor Lhx9; LIM/homeobox protein Lhx9; LIM-homeobox protein 9; LIM-homeodomain type transcription factor 9; si:dkey-121a9.2; zgc:112196 | |
Rabbit | |
Protein A | |
RUO | |
56956 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NQ69 | |
LHX9 | |
Synthetic peptide directed towards the C-terminal of human LHX9 (aa 324-373). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction