Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas™ Liat C. Diff Control Kit

Catalog No. 23312210 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-210 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-312-210 Supplier Roche Diagnostics cobas™ Supplier No. 07454970190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The cobas™ Liat C. difficile nucleic acid test for use on the cobas™ Liat System is a real-time PCR test that provides accurate results in 20 minutes for the detection of toxigenic C. diff in unformed stool stamples. This is for running a control test/lot validation for the assay.

  • The cobas™ Liat C. difficile nucleic acid test for use on the cobas™ Liat System is a real-time PCR test that provides accurate results in 20 minutes for the detection of toxigenic C. diff in unformed stool stamples
  • Provide clinician confidence with results in 20 minutes
  • Reduce enteric isolation time
  • Improves antibiotic stewardship initiatives by de-escalating days of unnecessary antibiotics
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets yes
Description Roche Diagnostics cobas™ Liat C.diff Control Kit
CE Marker Yes
Clia Complexity Moderate
Detection Method Target region is C. difficile Toxin B
Format Kit
Sample Type Unformed stool in cobas™ PCR media (used with Liat C.diff assay)
Sensitivity 93.5% (Liat C.diff assay)
Type Control Kit
Disposable Single-use
For Use With (Application) RT-PCR
For Use With (Equipment) Cobas Liat Analyzer
Includes One control Kit includes 5 positive and negative cobas Liat C.diff control sets
No. of Reactions 10 Control Sets
Packaging Type Kit
Quantity 1 Kit
Specificity 96.4% (Liat C.diff assay)
Analysis Time 20 min.
Dimensions (L x W x H) 5.28 x 2.64 x 2.48 in.
Weight (English) 0.18 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.