Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 125-2
Catalog No. M1501957140 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
60 SI
DIOL5
RP18
RP8
RPSELECTB
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase Mobile Phase Compatibility
M1501957140 RP8 RP (reversed-phase)
M1501957075 DIOL5 RP (when in mode) and NP
M1501957093 RPSELECTB RP (reversed-phase)
M1501957109 60 SI NP (normal-phase)
M1501957116 RP18 RP (reversed-phase)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M1501957140 Supplier MilliporeSigma™ Supplier No. 1501957140
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 125-2 comes with customized packing (please specify packing material) and is 125mmL x 2mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 2 mm
Length (Metric) 125 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase RP8
Packing Material Customized (specify packing material)
Column Type LiChroCART™ HPLC Cartridge