Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 250-3
Catalog No. M1501777075 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
60 SI
DIOL5
NH2
RP18
RP8
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase Mobile Phase Compatibility
M1501777075 DIOL5 RP (when in mode) and NP
M1501777076 NH2 RP (when in mode) and NP
M1501777109 60 SI NP (normal-phase)
M1501777139 RP8 RP (reversed-phase)
M1501777184 RP18 RP (reversed-phase)
M1501777174 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. M1501777075 Supplier MilliporeSigma™ Supplier No. 1501777075
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 250-3 comes with customized packing (please specify packing material) and is 250mmL x 3mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 3 mm
Length (Metric) 250 mm
Mobile Phase Compatibility RP (when in mode) and NP
Stationary Phase DIOL5
Packing Material Customized (specify packing material)
Column Type LiChroCART™ HPLC Cartridge